Skip to Content
Merck
All Photos(1)

Key Documents

L023050

Sigma-Aldrich

DCA Deblock

configured for (ÄKTA® and OligoPilot®)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116004

description

with Toluene

type

for DNA synthesis
for DNA synthesis

storage condition

dry at room temperature

concentration

42.2-51.6 g/L (DCA content, titration)

impurities

≤200 ppm water content (Karl Fischer)

color

clear

compatibility

configured for (ÄKTA® and OligoPilot®)

Looking for similar products? Visit Product Comparison Guide

General description

Deblock solution containing dichloroacetic acid (DCA) in toluene removes the dimethoxytrityl (DMT) protecting group from the 5′hydroxyl moiety of nucleotides already incorporated into the growing nucleic acid, prior to the addition of the next phosphoramidite. This solution contains toluene / Dichloroactic Acid (97/3 v/v).

Application

DCA Deblock solution facilitates removal of the DMT that allows the unprotected 5′hydroxyl moiety to react with a new phosphoramidite in a subsequent extension reaction.

Preparation Note

Toluene / Dichloroacetic acid 97/3 (v/v)

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Lact. - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service