Skip to Content
Merck
All Photos(2)

Key Documents

GE28-9269-73

HiScreen MabSelect

Cytiva 28-9269-73

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

recombinant protein A (E. coli)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9269-73

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

44 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning

2-12

working range

3-10

capacity

≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

General description

Determined at 10% breakthrough by frontal analysis at a mobile phase velocity of 500 cm/h in a column with a bed height of 20 cm.
HiScreen MabSelect is prepacked with MabSelect and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies (MAbs).
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Application

The small column volume of 4.7 mL and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

If necessary, two columns can easily be connected in series to give a bed height of 20 cm.

Media prepacked in HiScreen MabSelect columns are also available in other formats and as bulk packs, which makes it possible to use the same media for development work, pilot studies, and routine production.

Features and Benefits

  • MabSelect is a BioProcess medium for capture of MAbs from large sample volumes.
  • Prepacked, ready-to-use columns for convenient process development.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiScreen is a trademark of Cytiva
MabSelect is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service