ADF356301C-EP
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 Centriprep 100 - 125 mm, large rotor bore, pack of 6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
feature
holds 1 Centriprep 100 - 125 mm
packaging
pack of 6
manufacturer/tradename
Eppendorf® 022654331 (NA)
parameter
7,567 × g max. RCF
technique(s)
centrifugation: suitable
diam.
29.5 mm
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service