Skip to Content
Merck
All Photos(1)

Key Documents

SYNPLE-C001

Sigma-Aldrich

Reagent-cartridge Suzuki Coupling

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.22

General description

Reagent-cartridge Suzuki Coupling is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the synthesis of biaryl scaffolds through a Suzuki-Miyaura cross-coupling. The cartridge contains all necessary reagents and catalysts for coupling the user aryl bromide and aryl boronic acid.

Scale: up to 0.8 mmol
Compatible substrates: aryl bromides, aryl boronic acids

Cartridge contents:
  • Supported Pd catalyst, Cs2CO3, Celite (for cross-coupling reaction)
  • Supported carbonate (for purification)
  • Silica and HM-N (for purification)
Learn more about the Synple automated synthesis platform.

Explore all our boronic acids

Features and Benefits of Synple Platform
  • Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
  • Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety – all chemicals are contained in the system, minimizing the user’s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system – simple installation
  • Reaction temperatures from 25 °C – 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service