Recommended Products
grade
analytical standard
Quality Level
description
redox potential: 200 mV ±5 mV, 25°C
shelf life
limited shelf life, expiry date on the label
application(s)
environmental
pharmaceutical
format
mixture
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Broad range of values
- Very high specifications (±5mV)
- Detailed safety data sheet (SDS) available
- Certified with proven verifiable accuracy and uncertainty of measurement
- Guaranteed stability throughout its entire shelf life
Disclaimer

Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service