Skip to Content
Merck
All Photos(1)

Documents

TX1277

Supelco

Acetonitrile solution

suitable for HPLC, contains 0.1% (v/v) trifluoroacetic acid

Synonym(s):

Acetonitrile solution, ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide, acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
MDL number:
UNSPSC Code:
41116004
EC Index Number:
200-835-2

product name

Acetonitrile/TFA 0.1% Mobile Phase,

vapor pressure

97 hPa ( 20 °C)

Quality Level

form

liquid

concentration

0.095-0.105 % (v/v) in trifluoroacetic acid

technique(s)

HPLC: suitable

evapn. residue

≤5 ppm

color

APHA: ≤10

bp

81.6 °C/1,013 hPa

mp

-45.7 °C

transition temp

flash point 2 °C

density

0.782 g/mL at 25 °C

UV absorption

λ: 190 nm Amax: ≤1.00
λ: 200 nm Amax: ≤0.50
λ: 210 nm Amax: ≤0.40
λ: 230 nm Amax: ≤0.30
λ: 250 nm Amax: ≤0.05
λ: 280 nm Amax: ≤0.010
λ: 300 nm Amax: ≤0.010
λ: 350 nm Amax: ≤0.010

format

mixture

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TX1277P-1 is packaged in poly coated glass bottles

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (trifluoroacetic acid)0.095-0.105% (v/v)Color (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidGradient at 210 nm0.005AU maxGradient at 220 nm0.001AU maxGradient at 280 nm0.001AU maxUV Abs. at 190 nm1.00AU maxUV Abs. at 200 nm0.50AU maxUV Abs. at 210 nm0.40AU maxUV Abs. at 230 nm0.30AU maxUV Abs. at 250 nm0.05AU maxUV Abs. at 280 nm0.010AU maxUV Abs. at 300 nm0.010AU maxUV Abs. at 350 nm0.010AU max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service