Skip to Content
Merck
All Photos(1)

Key Documents

NA03

Sigma-Aldrich

Anti-PCNA (Ab-1) Mouse mAb (PC10)

liquid, clone PC10, Calbiochem®

Synonym(s):

Anti-Proliferating Cell Nuclear Antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PC10, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, yeast, mouse, human, zebrafish

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PCNA(5111)
zebrafish ... Pcna(30678)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/0-Ag14 mouse myeloma cells. Recognizes the ~37 kDa PCNA protein.
Recognizes the ~37 kDa PCNA protein in HeLa cells as well as in tonsil or normal colon tissue.
This Anti-PCNA (Ab-1) Mouse mAb (PC10) is validated for use in Immunoblotting, Immunofluorescence, Immunoprecipitation Paraffin Sections for the detection of PCNA (Ab-1).

Immunogen

Epitope: Within amino acids 112-121
recombinant PCNA

Application

Immunoblotting (2.5 µg/ml)

Immunofluorescence (1-5 µg/ml)

Immunoprecipitation (not recommended with unconjugated antibody, see comments)

Paraffin Sections (2.5 µg/ml, no pre-treatment required, see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Positive Control
HeLa cells, HL-60 cells, tonsil tissue, or normal colon tissue

Other Notes

Staining shows an intense nuclear pattern. Not recommended for immunoprecipitation because in some samples a ~37 kDa protein will be precipitated that is not PCNA. Generally, no pre-treatment is needed to stain paraffin sections, but staining may be enhanced in some samples by pretreating with 4 N HCl for 10 min at 37°C followed by 3 washes in water. Antibody should be titrated for optimal results in individual systems.
Waseem, N.H. and Lane, D.P. 1990. J. Cell. Sci.96, 121.
Suzuka, I., et al. 1989. Proc. Natl. Acad. Sci. USA86 3189.
Bravo, R., et al. 1987. Nature (London)326, 515.
Bravo, R. and MacDonald-Bravo, H. 1987. J. Cell. Biol.105, 1549.
Miyachi, K., et al. 1987. J. Immunol.121, 2228.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayu Inokuchi et al.
The Journal of experimental biology, 220(Pt 24), 4720-4732 (2017-11-01)
Spatiotemporal changes in branchial ionocyte distribution were investigated following transfer from seawater (SW) to freshwater (FW) in Japanese seabass. The mRNA expression levels of cystic fibrosis transmembrane conductance regulator (CFTR) and Na+/K+/2Cl- cotransporter 1a (NKCC1a) in the gills rapidly decreased
B K Law et al.
The Journal of biological chemistry, 275(49), 38261-38267 (2000-09-20)
Salicylate and its pro-drug form aspirin are widely used medicinally for their analgesic and anti-inflammatory properties, and more recently for their ability to protect against colon cancer and cardiovascular disease. Despite the wide use of salicylate, the mechanisms underlying its
Hailong Wang et al.
Journal of the American Heart Association, 8(14), e011994-e011994 (2019-07-13)
Background Exposure to chronic psychosocial stress is a risk factor for atherosclerosis-based cardiovascular disease. We previously demonstrated the increased expressions of cathepsin S (CatS) in atherosclerotic lesions. Whether CatS participates directly in stress-related neointimal hyperplasia has been unknown. Methods and
Alfeu Zanotto-Filho et al.
Cancer letters, 425, 101-115 (2018-04-03)
Molecular targeted compounds are emerging as a strategy to improve classical chemotherapy. Herein, we describe that using low dose of the multikinase inhibitor sorafenib improves cyclophosphamide antitumor activity by inhibiting angiogenesis, metastasis and promoting tumor healing in MDA-MB231 xenografts and
K Henke et al.
International journal of radiation oncology, biology, physics, 36(2), 385-391 (1996-09-01)
Stereotactic radiosurgery (RS) is being used increasingly for the treatment of small benign and malignant lesions, particularly in the brain. However, to fully realize the potential of this technique, more experimental data are needed. In this report we describe an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service