Skip to Content
Merck
All Photos(1)

Key Documents

MABF2105

Sigma-Aldrich

Anti-CD6 Antibody, clone UMCD6

clone UMCD6, from mouse

Synonym(s):

T-cell differentiation antigen CD6, T12, TP120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

UMCD6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

dot blot: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD6(923)

General description

T-cell differentiation antigen CD6 (UniProt: P30203; also known as T12, TP120, CD6) is encoded by the CD6 gene (Gene ID: 923) in human. CD6 is a single-pass type I membrane protein that serves as a cell adhesion molecule that mediates cell-cell contacts and regulates T-cell responses via its interaction with ALCAM/CD166. It contributes to signaling cascades triggered by activation of the TCR/CD3 complex and functions as a costimulatory molecule to promote T-cell activation and proliferation. It is also involved in the formation and maturation of the immunological synapse. CD6 has been detected on peripheral blood T-cells and on natural killer (NK) cells and is also detected in spleen, thymus, appendix, lymph nodes. CD6 is known to bind both lipopolysaccharide (LPS) from Gram-negative bacteria and lipoteichoic acid from Gram-positive bacteria and LPS binding leads to the activation of signaling cascades and down-stream MAP kinases. It is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off in the mature form. It has an extracellular domain (aa 18-402), a transmembrane domain (aa 403-423), and a cytoplasmic domain (aa 424-668). Seven isoforms of CD6 have been reported that are produced by alternative splicing.

Specificity

Clone UMCD6 specifically detects CD6 in human cells.

Immunogen

ST-1 T cells isolated from human knee synovium.

Application

Anti-CD6, clone UMCD6, Cat. No. MABF2105 is a mouse monoclonal antibody that detects CD6 and has been tested for use in Dot Blot, Flow Cytometry, Immunoprecipitation, Function Analysis, Inhibition assay.
Inhibition Analysis: A representative lot inhibited antigen-specific responses to tetanus toxoid, but not to the superantigen SEA. (Singer, N.G., et. al. (1996). Immunology. 88(4):537-43).

Induces Function: A representative lot showed strong synergistic effect with phorbol ester in inducing T cell activation and enhanced the autologous mixed lymphocyte reaction. (Bott, C.M., et. al. (1993). Int Immunol. 5(7):783-92).

Dot Blot Analysis: A representative lot detected CD6 in Dot Blot applications (Bott, C.M., et. al. (1993). Int Immunol. 5(7):783-92).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CD6 in Immunoprecipitation applications (Bott, C.M., et. al. (1993). Int Immunol. 5(7):783-92).

Flow Cytometry Analysis: A representative lot detected CD6 in Immunoprecipitation applications (Singer, N.G., et. al. (1996). Immunology. 88(4):537-43; Bott, C.M., et. al. (1993). Int Immunol. 5(7):783-92; Bott, C.M., et. al. (1994). J Immunol. 153(1):1-9; Li, Y., et. al. (2017). Proc Natl Acad Sci USA. 114(10):2687-2692).


Quality

Evaluated by Flow Cytometry in PMA stimulated Jurkat cells.

Flow Cytometry Analysis: A 1:1000 dilution of this antibody detected CD6 in one million Jurkat cells stimulated with PMA.

Target description

71.80 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service