8.41502
2,3-Dimethylmaleic anhydride
for synthesis
Synonym(s):
2,3-Dimethylmaleic anhydride, 3,4-Dimethyl-2,5-furandione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352115
EC Index Number:
212-165-8
NACRES:
NA.22
Quality Level
Assay
≥98.0% (GC)
form
crystals
mp
96 °C
density
1.107 g/cm3 at 100 °C
storage temp.
2-30°C
SMILES string
[o]1[c](c(c([c]1=O)C)C)=O
InChI
1S/C6H6O3/c1-3-4(2)6(8)9-5(3)7/h1-2H3
InChI key
MFGALGYVFGDXIX-UHFFFAOYSA-N
Analysis Note
Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 92 °C
Melting range (upper value): ≤ 95 °C
Identity (IR): passes test
Melting range (lower value): ≥ 92 °C
Melting range (upper value): ≤ 95 °C
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service