Skip to Content
Merck
All Photos(1)

Key Documents

182750

Millipore

ASB-14

A zwitterionic amidosulfobetaine detergent useful for solubilizing proteins visualized by 2D-electrophoresis.

Synonym(s):

ASB-14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46N2O4S
Molecular Weight:
434.68
UNSPSC Code:
12161902
NACRES:
NA.21

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

10-30°C

General description

A zwitterionic amidosulfobetaine detergent that exhibits better protein extraction properties than CHAPS.
A zwitterionic amidosulfobetaine detergent useful for solubilizing proteins visualized by 2D-electrophoresis. The increased solubilization produced by this detergent enables the identification of previously undetected membrane proteins.

Application


  • Effect of bovine pericardial extracellular matrix scaffold niche on seeded human mesenchymal stem cell function.: This paper explores the influence of the bovine pericardial extracellular matrix on human mesenchymal stem cells, with ASB-14 used to modulate cellular responses. The research contributes to advancements in scaffold-based tissue engineering (Liu et al., 2016).

  • In vivo xenogeneic scaffold fate is determined by residual antigenicity and extracellular matrix preservation.: Investigating the fate of xenogeneic scaffolds in vivo, this study shows that ASB-14 helps maintain extracellular matrix integrity and reduce antigenicity, which is critical for successful scaffold integration and function in tissue engineering (Wong et al., 2016).

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Carroll, J., et al. J. Biol. Chem.277, 50311.
Herbert, B. 1999. Electrophoresis20, 660.
Chevallet, M. et al. 1998. Electrophoresis 19, 1901.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service