Key Documents
Z673579
LAB-AIRE® II electrical benchtop dryers
AC/DC input 110 V AC, single sided, 3 tier
About This Item
UNSPSC Code:
41103200
feature
CSA approved: no
no. of pegs 38
manufacturer/tradename
Bel-Art F188190013
L × W × H
16 in. × 7.5 in. × 23.75 in.
General description
LAB-AIRE II electrical benchtop dryers provide continuous warm air circulation for quick drying of labware. Selector knob allows choice of 3 temperature settings: high, medium and low. The warmed air is kept dust free with a replaceable polyurethane foam filter. Airflow can be redirected to only the pegs in use by replacing the unused pegs with plugs (6 plugs included).
Select from single and double-sided models with up to 76 peg locations to hold glass or plastic labware such as beakers, flasks, bottles, test tubes and cylinders. Removable polypropylene baskets for bulk rinsing, drying, transport and storage of small labware and extra pegs are available separately.
  • Small, compact footprint
  • Single sided models can be placed against a wall conserving bench space
  • High capacity, double-sided models provide twice capacity with only slightly larger footprint
  • Filtered warm aire keeps labware dust free
Legal Information
Lab-Aire is a registered trademark of SP Scienceware, a division of SP