Key Documents
Z505846
Sigma-Aldrich
Nalgene® quick filling/venting closure
number of ports, 2, fits tubing, 3/8 in.
Synonym(s):
transfer cap, vented cap
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
feature
fits tubing 3/8 in.
number of ports 2
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 2158-0022
General description
Available in two and three port configurations, and is ideal for filling/dispensing into and out of vacuum trap systems with carboys and bottles. Quick disconnect ports fit either 1/4 in. or 3/8 in. I.D. tubing, and have a built-in shut off valve if tubing inserts are taken out. Fits onto all carboys and bottles with 83 mm neck. Leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries