905488
N-Acetyl-α-D-15N glucosamine-1-phosphate disodium salt
≥98 atom % 15N, ≥95% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H1415NNa2O9P
Molecular Weight:
346.14
UNSPSC Code:
12352116
Recommended Products
isotopic purity
≥98 atom % 15N
Assay
≥95% (CP)
form
solid
mass shift
M+1
storage temp.
−20°C
SMILES string
O[C@@H]1[C@H](O[C@H](OP(O)(O)=O)[C@H]([15NH]C(C)=O)[C@H]1O)CO
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service