Skip to Content
Merck
All Photos(1)

Key Documents

197718

Sigma-Aldrich

3-Methyl-2-cyclohexen-1-ol

96%

Synonym(s):

(±)-3-Hydroxy-1-methylcyclohexene, (±)-3-Methyl-2-cyclohexen-1-ol, (±)-Seudenol, 1-Methyl-1-cyclohexen-3-ol, 1-Methyl-3-hydroxycyclohex-1-ene, 3-Hydroxy-1-methylcyclohexene, 3-Methyl-2-cyclohexen-1-ol, 3-Methyl-2-cyclohexenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H8OH
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.484 (lit.)

bp

56 °C/1 mmHg (lit.)

density

0.946 g/mL at 25 °C (lit.)

SMILES string

CC1=CC(O)CCC1

InChI

1S/C7H12O/c1-6-3-2-4-7(8)5-6/h5,7-8H,2-4H2,1H3

InChI key

XNDZQQSKSQTQQD-UHFFFAOYSA-N

Application

3-Methyl-2-cyclohexen-1-ol was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

X Zhou et al.
Journal of medicinal chemistry, 42(18), 3539-3556 (1999-09-10)
The synthesis of four CD-ring-modified 19-nor-1alpha, 25-dihydroxyvitamin D(3) derivatives lacking C15, referred to as 6C analogues, and diastereomeric at C17 and C20 is described. The synthesis involves an Ireland-Claisen rearrangement of a 3-methyl-substituted ester of (1R)-3-methyl-2-cyclohexen-1-ol as the key step
Aldrichimica Acta, 16, 41-41 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service