Skip to Content
Merck
All Photos(1)

Key Documents

115851

Sigma-Aldrich

2-Bromo-1,3-dimethylbenzene

98%

Synonym(s):

2-Bromo-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3Br
CAS Number:
Molecular Weight:
185.06
Beilstein:
1929780
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.555 (lit.)

bp

206 °C (lit.)

mp

−10 °C (lit.)

density

1.389 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)c1Br

InChI

1S/C8H9Br/c1-6-4-3-5-7(2)8(6)9/h3-5H,1-2H3

InChI key

MYMYVYZLMUEVED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Bromo-1,3-dimethylbenzene has been used to prepare 2,2′,4,6,6′-pentamethylbiphenyl.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Schmidt et al.
Chemical communications (Cambridge, England), 46(17), 2995-2997 (2010-04-14)
The catalyst system consisting of 3,3'-(3,4-bis(dichloro-methylene)cyclobut-1-ene-1,2-diyl)bis(1-methyl-1H-imidazolium) bis(tetrafluoroborate), Pd(OAc)(2) and NaOtBu in toluene proved to be very effective for a broad variety of Suzuki-Miyaura reactions at room temperature. It is also suited for the synthesis of sterically hindered compounds including 2,6-di-tert-butyl-2'-substituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service