Merck
  • Concise review: transplantation of cultured oral mucosal epithelial cells for treating limbal stem cell deficiency-current status and future perspectives.
Concise review: transplantation of cultured oral mucosal epithelial cells for treating limbal stem cell deficiency-current status and future perspectives.
Stem cells (Dayton, Ohio) (2015-03-20)
Tor Paaske Utheim
RESUMEN
A number of diseases and external factors can deplete limbal stem cells, causing pain and visual loss. Ten years have passed since the first transplantation of cultured oral mucosal epithelial cells in humans, representing the first autologous cell-based therapy for severe bilateral limbal stem cell deficiency. Its steady increase in popularity since then can be attributed to the accumulating evidence of its efficacy in reverting limbal stem cell deficiency. In this review, the focus is on clinical, and to a lesser degree laboratory, features of cultured oral mucosal epithelial transplants over the past 10 years. Comparisons with other available technologies are made. Avenues for research to stimulate further improvements in clinical results and allow worldwide distribution of limbal stem cell therapy based on oral mucosal cells are discussed. These include storage and transportation of cultured oral mucosal epithelial sheets and in vivo culture of oral mucosal epithelial cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%