Merck
  • MBX-102/JNJ39659100, a novel peroxisome proliferator-activated receptor-ligand with weak transactivation activity retains antidiabetic properties in the absence of weight gain and edema.
MBX-102/JNJ39659100, a novel peroxisome proliferator-activated receptor-ligand with weak transactivation activity retains antidiabetic properties in the absence of weight gain and edema.
Molecular endocrinology (Baltimore, Md.) (2009-04-25)
Francine M Gregoire, Fang Zhang, Holly J Clarke, Thomas A Gustafson, Dorothy D Sears, Svetlana Favelyukis, James Lenhard, Dennis Rentzeperis, L Edward Clemens, Yi Mu, Brian E Lavan
ABSTRACT
MBX-102/JNJ39659100 (MBX-102) is in clinical development as an oral glucose-lowering agent for the treatment of type 2 diabetes. MBX-102 is a nonthiazolidinedione (TZD) selective partial agonist of peroxisome proliferator-activated receptor (PPAR)-gamma that is differentiated from the TZDs structurally, mechanistically, preclinically and clinically. In diabetic rodent models, MBX-102 has insulin-sensitizing and glucose-lowering properties comparable to TZDs without dose-dependent increases in body weight. In vitro, in contrast with full PPAR-gamma agonist treatment, MBX-102 fails to drive human and murine adipocyte differentiation and selectively modulates the expression of a subset of PPAR-gamma target genes in mature adipocytes. Moreover, MBX-102 does not inhibit osteoblastogenesis of murine mesenchymal cells. Compared with full PPAR-gamma agonists, MBX-102 displays differential interactions with the PPAR-gamma ligand binding domain and possesses reduced ability to recruit coactivators. Interestingly, in primary mouse macrophages, MBX-102 displays enhanced antiinflammatory properties compared with other PPAR-gamma or alpha/gamma agonists, suggesting that MBX-102 has more potent transrepression activity. In summary, MBX-102 is a selective PPAR-gamma modulator with weak transactivation but robust transrepression activity. MBX-102 exhibits full therapeutic activity without the classical PPAR-gamma side effects and may represent the next generation insulin sensitizer.
Sigma-Aldrich
MBX-102, ≥98% (HPLC)
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
GW7647, ≥98% (HPLC)