Merck
  • A Small Molecule That Protects the Integrity of the Electron Transfer Chain Blocks the Mitochondrial Apoptotic Pathway.
A Small Molecule That Protects the Integrity of the Electron Transfer Chain Blocks the Mitochondrial Apoptotic Pathway.
Molecular cell (2016-07-23)
Xian Jiang, Li Li, Zhengxin Ying, Chenjie Pan, Shaoqiang Huang, Lin Li, Miaomiao Dai, Bo Yan, Ming Li, Hui Jiang, She Chen, Zhiyuan Zhang, Xiaodong Wang
ABSTRACT
In response to apoptotic stimuli, mitochondria in mammalian cells release cytochrome c and other apoptogenic proteins, leading to the subsequent activation of caspases and apoptotic cell death. This process is promoted by the pro-apoptotic members of the Bcl-2 family of proteins, such as Bim and Bax, which, respectively, initiate and execute cytochrome c release from the mitochondria. Here we report the discovery of a small molecule that efficiently blocks Bim-induced apoptosis after Bax is activated on the mitochondria. The cellular target of this small molecule was identified to be the succinate dehydrogenase subunit B (SDHB) protein of complex II of the mitochondrial electron transfer chain (ETC). The molecule protects the integrity of the ETC and allows treated cells to continue to proliferate after apoptosis induction. Moreover, this molecule blocked dopaminergic neuron death and reversed Parkinson-like behavior in a rat model of Parkinson's disease.
Sigma-Aldrich
Monoclonal Anti-ATG5 antibody produced in mouse, clone ATG5-18, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.