Merck
  • Nitric oxide, afferent sensory nerves, and antioxidative enzymes in the mechanism of protection mediated by tricarbonyldichlororuthenium(II) dimer and sodium hydrosulfide against aspirin-induced gastric damage.
Nitric oxide, afferent sensory nerves, and antioxidative enzymes in the mechanism of protection mediated by tricarbonyldichlororuthenium(II) dimer and sodium hydrosulfide against aspirin-induced gastric damage.
Journal of gastroenterology (2017-02-27)
Marcin Magierowski, Magdalena Hubalewska-Mazgaj, Katarzyna Magierowska, Dagmara Wojcik, Zbigniew Sliwowski, Slawomir Kwiecien, Tomasz Brzozowski
ABSTRACT
Aspirin exerts side effects within the gastrointestinal tract. Hydrogen sulfide (H Wistar rats with or without capsaicin-induced denervation of sensory neurons were pretreated with vehicle, CORM-2 (5 mg/kg intragastrically), or NaHS (5 mg/kg intragastrically) with or without capsazepine (5 mg/kg intragastrically) or N Aspirin caused gastric lesions, decreased GBF, and raised MDA content, but pretreatment with NaHS and CORM-2 reduced these effects. Capsaicin-induced denervation or co-treatment with capsazepine reversed the gastroprotective and vasodilatory effects of NaHS but not those of CORM-2. L-NNA reversed NaHS-induced gastroprotection and partly reduced CORM-2-induced gastroprotection. NaHS and CORM-2 decreased MDA and 4-HNE content, restoring GPx-1 protein expression. We conclude that H
Sigma-Aldrich
1-Methyl-2-phenylindole, 99%