Merck
  • PEBP1 Wardens Ferroptosis by Enabling Lipoxygenase Generation of Lipid Death Signals.
PEBP1 Wardens Ferroptosis by Enabling Lipoxygenase Generation of Lipid Death Signals.
Cell (2017-10-21)
Sally E Wenzel, Yulia Y Tyurina, Jinming Zhao, Claudette M St Croix, Haider H Dar, Gaowei Mao, Vladimir A Tyurin, Tamil S Anthonymuthu, Alexandr A Kapralov, Andrew A Amoscato, Karolina Mikulska-Ruminska, Indira H Shrivastava, Elizabeth M Kenny, Qin Yang, Joel C Rosenbaum, Louis J Sparvero, David R Emlet, Xiaoyan Wen, Yoshinori Minami, Feng Qu, Simon C Watkins, Theodore R Holman, Andrew P VanDemark, John A Kellum, Ivet Bahar, Hülya Bayır, Valerian E Kagan
ABSTRACT
Ferroptosis is a form of programmed cell death that is pathogenic to several acute and chronic diseases and executed via oxygenation of polyunsaturated phosphatidylethanolamines (PE) by 15-lipoxygenases (15-LO) that normally use free polyunsaturated fatty acids as substrates. Mechanisms of the altered 15-LO substrate specificity are enigmatic. We sought a common ferroptosis regulator for 15LO. We discovered that PEBP1, a scaffold protein inhibitor of protein kinase cascades, complexes with two 15LO isoforms, 15LO1 and 15LO2, and changes their substrate competence to generate hydroperoxy-PE. Inadequate reduction of hydroperoxy-PE due to insufficiency or dysfunction of a selenoperoxidase, GPX4, leads to ferroptosis. We demonstrated the importance of PEBP1-dependent regulatory mechanisms of ferroptotic death in airway epithelial cells in asthma, kidney epithelial cells in renal failure, and cortical and hippocampal neurons in brain trauma. As master regulators of ferroptotic cell death with profound implications for human disease, PEBP1/15LO complexes represent a new target for drug discovery.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Locostatin, ≥98% (HPLC)
Sigma-Aldrich
ML351, ≥98% (HPLC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)