- A Stat6/Pten Axis Links Regulatory T Cells with Adipose Tissue Function.
A Stat6/Pten Axis Links Regulatory T Cells with Adipose Tissue Function.
Cell metabolism (2017-09-07)
Stefanie Kälin, Maike Becker, Verena B Ott, Isabelle Serr, Fabian Hosp, Mohammad M H Mollah, Susanne Keipert, Daniel Lamp, Francoise Rohner-Jeanrenaud, Victoria K Flynn, Martin G Scherm, Lucas F R Nascimento, Katharina Gerlach, Vanessa Popp, Sarah Dietzen, Tobias Bopp, Purna Krishnamurthy, Mark H Kaplan, Manuel Serrano, Stephen C Woods, Philipp Tripal, Ralf Palmisano, Martin Jastroch, Matthias Blüher, Christian Wolfrum, Benno Weigmann, Anette-Gabriele Ziegler, Matthias Mann, Matthias H Tschöp, Carolin Daniel
PMID28877454