Merck
  • Novel mononuclear Cu (II) terpyridine complexes: Impact of fused ring thiophene and thiazole head groups towards DNA/BSA interaction, cleavage and antiproliferative activity on HepG2 and triple negative CAL-51 cell line.
Novel mononuclear Cu (II) terpyridine complexes: Impact of fused ring thiophene and thiazole head groups towards DNA/BSA interaction, cleavage and antiproliferative activity on HepG2 and triple negative CAL-51 cell line.
European journal of medicinal chemistry (2017-05-06)
V M Manikandamathavan, M Thangaraj, T Weyhermuller, R P Parameswari, V Punitha, N Narasimha Murthy, Balachandran Unni Nair
ABSTRACT
Two mononuclear copper (II) terpyridine complexes namely, [Cu(Btptpy) (ClO
Sigma-Aldrich
Benzo[b]thiophene-2-carboxaldehyde, 97%
Sigma-Aldrich
Benzothiazole-2-carboxaldehyde, 97%
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)