Merck
  • Clinicopathological examination of dipeptidase 1 expression in colorectal cancer.
Clinicopathological examination of dipeptidase 1 expression in colorectal cancer.
Biomedical reports (2017-04-18)
Kazunoshin Tachibana, Motonobu Saito, Jun-Ichi Imai, Emi Ito, Yuka Yanagisawa, Reiko Honma, Katsuharu Saito, Jin Ando, Tomoyuki Momma, Shinji Ohki, Tohru Ohtake, Shinya Watanabe, Satoshi Waguri, Seiichi Takenoshita
ABSTRACT
Dipeptidase 1 (DPEP1) is a zinc-dependent metalloproteinase that is fundamental in glutathione and leukotriene metabolism.
Sigma-Aldrich
Anti-DPEP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available. Contact Technical Service