Merck
  • Pharmacokinetic properties of the synthetic cannabinoid JWH-018 and of its metabolites in serum after inhalation.
Pharmacokinetic properties of the synthetic cannabinoid JWH-018 and of its metabolites in serum after inhalation.
Journal of pharmaceutical and biomedical analysis (2017-04-04)
Stefan W Toennes, Anna Geraths, Werner Pogoda, Alexander Paulke, Cora Wunder, Eef L Theunissen, Johannes G Ramaekers
ABSTRACT
Each year, synthetic cannabinoids are occurring in high numbers in the illicit drug market, but data on their pharmacology and toxicology are scarcely available. Therefore, a pilot study was performed to assess adverse effects of JWH-018, which is one of the oldest and best known synthetic cannabinoids. Six subjects inhaled smoke from 2 and 3mg JWH-018. The drug and nine of its metabolites were analyzed in their blood samples taken during the following 12h by liquid chromatography-mass spectrometry (LC-MSMS). The maximum concentration of JWH-018 reached 2.9-9.9ng/ml after inhalation and markedly decreased during the next 1.5h, followed by a multiexponential decline (t
Sigma-Aldrich
4-Hydroxyindole, 99%