Merck
  • Evidence for glutamate as a neuroglial transmitter within sensory ganglia.
Evidence for glutamate as a neuroglial transmitter within sensory ganglia.
PloS one (2013-07-12)
Ling-Hsuan Kung, Kerui Gong, Mary Adedoyin, Johnson Ng, Aditi Bhargava, Peter T Ohara, Luc Jasmin
ABSTRACT
This study examines key elements of glutamatergic transmission within sensory ganglia of the rat. We show that the soma of primary sensory neurons release glutamate when depolarized. Using acute dissociated mixed neuronal/glia cultures of dorsal root ganglia (DRG) or trigeminal ganglia and a colorimetric assay, we show that when glutamate uptake by satellite glial cells (SGCs) is inhibited, KCl stimulation leads to simultaneous increase of glutamate in the culture medium. With calcium imaging we see that the soma of primary sensory neurons and SGCs respond to AMPA, NMDA, kainate and mGluR agonists, and selective antagonists block this response. Using whole cell patch-clamp technique, inward currents were recorded from small diameter (<30 µm) DRG neurons from intact DRGs (ex-vivo whole ganglion preparation) in response to local application of the above glutamate receptor agonists. Following a chronic constriction injury (CCI) of either the inferior orbital nerve or the sciatic nerve, glutamate expression increases in the trigeminal ganglia and DRG respectively. This increase occurs in neurons of all diameters and is present in the somata of neurons with injured axons as well as in somata of neighboring uninjured neurons. These data provides additional evidence that glutamate can be released within the sensory ganglion, and that the somata of primary sensory neurons as well as SGCs express functional glutamate receptors at their surface. These findings, together with our previous gene knockdown data, suggest that glutamatergic transmission within the ganglion could impact nociceptive threshold.
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-NMDAR1 Antibody, rabbit monoclonal, culture supernatant, clone 1.17.2.6, Chemicon®
Sigma-Aldrich
Anti-Glutamate Receptor 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutamine Synthetase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution