Merck
  • OCIAD2 activates γ-secretase to enhance amyloid β production by interacting with nicastrin.
OCIAD2 activates γ-secretase to enhance amyloid β production by interacting with nicastrin.
Cellular and molecular life sciences : CMLS (2013-11-26)
Jonghee Han, Sunmin Jung, Jiyeon Jang, Tae-In Kam, Hyunwoo Choi, Byung-Ju Kim, Jihoon Nah, Dong-Gyu Jo, Toshiyuki Nakagawa, Masaki Nishimura, Yong-Keun Jung
ABSTRACT
The gamma (γ)-secretase holoenzyme is composed of four core proteins and cleaves APP to generate amyloid beta (Aβ), a key molecule that causes major neurotoxicity during the early stage of Alzheimer's disease (AD). However, despite its important role in Aβ production, little is known about the regulation of γ-secretase. OCIAD2, a novel modulator of γ-secretase that stimulates Aβ production, and which was isolated from a genome-wide functional screen using cell-based assays and a cDNA library comprising 6,178 genes. Ectopic expression of OCIAD2 enhanced Aβ production, while reduction of OCIAD2 expression suppressed it. OCIAD2 expression facilitated the formation of an active γ-secretase complex and enhanced subcellular localization of the enzyme components to lipid rafts. OCIAD2 interacted with nicastrin to stimulate γ-secretase activity. OCIAD2 also increased the interaction of nicastrin with C99 and stimulated APP processing via γ-secretase activation, but did not affect Notch processing. In addition, a cell-permeable Tat-OCIAD2 peptide that interfered with the interaction of OCIAD2 with nicastrin interrupted the γ-secretase-mediated AICD production. Finally, OCIAD2 expression was significantly elevated in the brain of AD patients and PDAPP mice. This study identifies OCIAD2 as a selective activator of γ-secretase to increase Aβ generation.
Sigma-Aldrich
Anti-OCIAD2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution, ab2
Sigma-Aldrich
Anti-ADAM 10 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution