Merck
  • On the Mesoporogen-Free Synthesis of Single-Crystalline Hierarchically Structured ZSM-5 Zeolites in a Quasi-Solid-State System.
On the Mesoporogen-Free Synthesis of Single-Crystalline Hierarchically Structured ZSM-5 Zeolites in a Quasi-Solid-State System.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-04-24)
Tongguang Ge, Zile Hua, Xiaoyun He, Jian Lv, Hangrong Chen, Lingxia Zhang, Heliang Yao, Ziwei Liu, Chucheng Lin, Jianlin Shi
ABSTRACT
Hierarchically structured zeolites (HSZs) have gained much academic and industrial interest owing to their multiscale pore structures and consequent excellent performances in varied chemical processes. Although a number of synthetic strategies have been developed in recent years, the scalable production of HSZs single crystals with penetrating and three-dimensionally (3-D) interconnected mesopore systems but without using a mesoscale template is still a great challenge. Herein, based on a steam-assisted crystallization (SAC) method, we report a facile and scalable strategy for the synthesis of single-crystalline ZSM-5 HSZs by using only a small amount of micropore-structure-directing agents (i.e., tetrapropylammonium hydroxide). The synthesized materials exhibited high crystallinity, a large specific surface area of 468 m(2)  g(-1) , and a pore volume of 0.43 cm(3)  g(-1) without sacrificing the microporosity (≈0.11 cm(3)  g(-1) ) in a product batch up to 11.7 g. Further, a kinetically controlled nucleation-growth mechanism is proposed for the successful synthesis of single-crystalline ZSM-5 HSZs with this novel process. As expected, compared with the conventional microporous ZSM-5 and amorphous mesoporous Al-MCM-41 counterparts, the synthesized HSZs exhibited significantly enhanced activity and stability and prolonged lifetime in model reactions, especially when bulky molecules were involved.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Pricing and availability is not currently available.
Sigma-Aldrich
1,4-Diisopropylbenzene, 97%
Pricing and availability is not currently available.