Merck
  • Mitochondrial uncouplers inhibit clathrin-mediated endocytosis largely through cytoplasmic acidification.
Mitochondrial uncouplers inhibit clathrin-mediated endocytosis largely through cytoplasmic acidification.
Nature communications (2016-06-09)
Wim Dejonghe, Sabine Kuenen, Evelien Mylle, Mina Vasileva, Olivier Keech, Corrado Viotti, Jef Swerts, Matyáš Fendrych, Fausto Andres Ortiz-Morea, Kiril Mishev, Simon Delang, Stefan Scholl, Xavier Zarza, Mareike Heilmann, Jiorgos Kourelis, Jaroslaw Kasprowicz, Le Son Long Nguyen, Andrzej Drozdzecki, Isabelle Van Houtte, Anna-Mária Szatmári, Mateusz Majda, Gary Baisa, Sebastian York Bednarek, Stéphanie Robert, Dominique Audenaert, Christa Testerink, Teun Munnik, Daniël Van Damme, Ingo Heilmann, Karin Schumacher, Johan Winne, Jiří Friml, Patrik Verstreken, Eugenia Russinova
ABSTRACT
ATP production requires the establishment of an electrochemical proton gradient across the inner mitochondrial membrane. Mitochondrial uncouplers dissipate this proton gradient and disrupt numerous cellular processes, including vesicular trafficking, mainly through energy depletion. Here we show that Endosidin9 (ES9), a novel mitochondrial uncoupler, is a potent inhibitor of clathrin-mediated endocytosis (CME) in different systems and that ES9 induces inhibition of CME not because of its effect on cellular ATP, but rather due to its protonophore activity that leads to cytoplasm acidification. We show that the known tyrosine kinase inhibitor tyrphostinA23, which is routinely used to block CME, displays similar properties, thus questioning its use as a specific inhibitor of cargo recognition by the AP-2 adaptor complex via tyrosine motif-based endocytosis signals. Furthermore, we show that cytoplasm acidification dramatically affects the dynamics and recruitment of clathrin and associated adaptors, and leads to reduction of phosphatidylinositol 4,5-biphosphate from the plasma membrane.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Endosidin9, ≥98% (HPLC)