Merck
  • Hepatocyte growth factor activator inhibitor type 1 maintains the assembly of keratin into desmosomes in keratinocytes by regulating protease-activated receptor 2-dependent p38 signaling.
Hepatocyte growth factor activator inhibitor type 1 maintains the assembly of keratin into desmosomes in keratinocytes by regulating protease-activated receptor 2-dependent p38 signaling.
The American journal of pathology (2015-04-07)
Makiko Kawaguchi, Ai Kanemaru, Akira Sawaguchi, Koji Yamamoto, Takashi Baba, Chen-Yong Lin, Michael D Johnson, Tsuyoshi Fukushima, Hiroaki Kataoka
ABSTRACT
Hepatocyte growth factor activator inhibitor type 1 (HAI-1; official symbol SPINT1) is a membrane-associated serine proteinase inhibitor abundantly expressed in epithelial tissues. Genetically engineered mouse models demonstrated that HAI-1 is critical for epidermal function, possibly through direct and indirect regulation of cell surface proteases, such as matriptase and prostasin. To obtain a better understanding of the role of HAI-1 in maintaining epidermal integrity, we performed ultrastructural analysis of Spint1-deleted mouse epidermis and organotypic culture of an HAI-1 knockdown (KD) human keratinocyte cell line, HaCaT. We found that the aggregation of tonofilaments to desmosomes was significantly reduced in HAI-1-deficient mouse epidermis with decreased desmosome number. Similar findings were observed in HAI-1 KD HaCaT organotypic cultures. Immunoblot and immunohistochemical analyses revealed that p38 mitogen-activated protein kinase was activated in response to HAI-1 insufficiency. Treatment of HAI-1 KD HaCaT cells with a p38 inhibitor abrogated the above-observed ultrastructural abnormalities. The activation of p38 induced by the loss of HAI-1 likely resulted from enhanced signaling of protease-activated receptor-2 (PAR-2), because its silencing abrogated the enhanced activation of p38. Consequently, treatment of HAI-1 KD HaCaT cells with a serine protease inhibitor, aprotinin, or PAR-2 antagonist alleviated the abnormal ultrastructural phenotype in organotypic culture. These results suggest that HAI-1 may have a critical role in maintaining normal keratinocyte morphology through regulation of PAR-2-dependent p38 mitogen-activated protein kinase signaling.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
HEPES
Sigma-Aldrich
MISSION® esiRNA, targeting human ST14
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets