Merck
  • Effects of water-saving irrigation on emissions of greenhouse gases and prokaryotic communities in rice paddy soil.
Effects of water-saving irrigation on emissions of greenhouse gases and prokaryotic communities in rice paddy soil.
Microbial ecology (2014-04-01)
Jae-Hyung Ahn, Min-Young Choi, Byung-Yong Kim, Jong-Sik Lee, Jaekyeong Song, Gun-Yeob Kim, Hang-Yeon Weon
ABSTRACT
The effects of water-saving irrigation on emissions of greenhouse gases and soil prokaryotic communities were investigated in an experimental rice field. The water layer was kept at 1-2 cm in the water-saving (WS) irrigation treatment and at 6 cm in the continuous flooding (CF) irrigation treatment. WS irrigation decreased CH(4) emissions by 78 % and increased N(2)O emissions by 533 %, resulting in 78 % reduction of global warming potential compared to the CF irrigation. WS irrigation did not affect the abundance or phylogenetic distribution of bacterial/archaeal 16S rRNA genes and the abundance of bacterial/archaeal 16S rRNAs. The transcript abundance of CH(4) emission-related genes generally followed CH(4) emission patterns, but the difference in abundance between mcrA transcripts and amoA/pmoA transcripts best described the differences in CH(4) emissions between the two irrigation practices. WS irrigation increased the relative abundance of 16S rRNAs and functional gene transcripts associated with Anaeromyxobacter and Methylocystis spp., suggesting that their activities might be important in emissions of the greenhouse gases. The N(2)O emission patterns were not reflected in the abundance of N(2)O emission-related genes and transcripts. We showed that the alternative irrigation practice was effective for mitigating greenhouse gas emissions from rice fields and that it did not affect the overall size and structure of the soil prokaryotic community but did affect the activity of some groups.
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%