Merck
  • Rituximab as induction therapy after renal transplantation: a randomized, double-blind, placebo-controlled study of efficacy and safety.
Rituximab as induction therapy after renal transplantation: a randomized, double-blind, placebo-controlled study of efficacy and safety.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2015-01-24)
M W F van den Hoogen, E G Kamburova, M C Baas, E J Steenbergen, S Florquin, H J P M Koenen, I Joosten, L B Hilbrands
ABSTRACT
We evaluated the efficacy and safety of rituximab as induction therapy in renal transplant patients. In a double-blind, placebo-controlled study, 280 adult renal transplant patients were randomized between a single dose of rituximab (375 mg/m(2)) or placebo during transplant surgery. Patients were stratified according to panel-reactive antibody (PRA) value and rank number of transplantation. Maintenance immunosuppression consisted of tacrolimus, mycophenolate mofetil and steroids. The primary endpoint was the incidence of biopsy proven acute rejection (BPAR) within 6 months after transplantation. The incidence of BPAR was comparable between rituximab-treated (23/138, 16.7%) and placebo-treated patients (30/142, 21.2%, p = 0.25). Immunologically high-risk patients (PRA >6% or re-transplant) not receiving rituximab had a significantly higher incidence of rejection (13/34, 38.2%) compared to other treatment groups (rituximab-treated immunologically high-risk patients, and rituximab- or placebo-treated immunologically low-risk (PRA ≤ 6% or first transplant) patients (17.9%, 16.4% and 15.7%, p = 0.004). Neutropenia (<1.5 × 10(9) /L) occurred more frequently in rituximab-treated patients (24.3% vs. 2.2%, p < 0.001). After 24 months, the cumulative incidence of infections and malignancies was comparable. A single dose of rituximab as induction therapy did not reduce the overall incidence of BPAR, but might be beneficial in immunologically high-risk patients. Treatment with rituximab was safe.
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
USP
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
Supelco
Mycophenolate Mofetil, Pharmaceutical Secondary Standard; Certified Reference Material
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard