Merck
  • Discovery and characterization of terpenoid biosynthetic pathways of fungi.
Discovery and characterization of terpenoid biosynthetic pathways of fungi.
Methods in enzymology (2012-09-25)
Grayson T Wawrzyn, Sarah E Bloch, Claudia Schmidt-Dannert
ABSTRACT
Fungi produce a myriad of terpenoids with a broad range of biological activities, many of which can be adapted to human use. This requires knowledge of the enzymes responsible for the biosynthesis of these compounds. Herein, we describe strategies for identification and characterization of putative biosynthetic genes, structural examination of important pathway enzymes with a focus on altering activity, and identification of biosynthetic clusters, and genome mining for yet-to-be-discovered pathways. Fungi are a particularly attractive class of organism for terpenoid pathway discovery, as they often cluster their biosynthetic genes. The affordability of genome sequencing and the relatively small size of fungal genomes further simplify this process. While only a select few fungal strains are genetically tractable, many terpenoid biosynthetic genes are functional in Escherichia coli and Saccharomyces cerevisiae, allowing easy characterization. Identification of new terpenoid biosynthetic pathways has the potential to uncover new pharmaceutical compounds and establish new strategies for metabolic engineering.
Product Description
Supelco
Supelpak-2SV, matrix styrene-divinylbenzene, 20-60 mesh, pkg of 100 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 mg
Fulfilment and delivery delayed
100 mg
Fulfilment and delivery delayed
500 mg
Fulfilment and delivery delayed
1 g
Fulfilment and delivery delayed
To order products, please contact your local dealer.
Supelco
Supelpak-2SV, matrix styrene-divinylbenzene, 20-60 mesh, pkg of 1000 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 mg
Fulfilment and delivery delayed
100 mg
Fulfilment and delivery delayed
500 mg
Fulfilment and delivery delayed
1 g
Fulfilment and delivery delayed
To order products, please contact your local dealer.
Supelco
Supelpak-2SV, matrix styrene-divinylbenzene, 20-60 mesh, pkg of 250 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 mg
Fulfilment and delivery delayed
100 mg
Fulfilment and delivery delayed
500 mg
Fulfilment and delivery delayed
1 g
Fulfilment and delivery delayed
To order products, please contact your local dealer.