Merck
  • Further field evaluation of synthetic herbivore-induced plant volatiles as attractants for beneficial insects.
Further field evaluation of synthetic herbivore-induced plant volatiles as attractants for beneficial insects.
Journal of chemical ecology (2005-05-19)
David G James
ABSTRACT
Fifteen synthetic herbivore-induced plant volatiles (HIPVs) were field-tested for attractivity to beneficial insects in two experiments conducted in an open field and a hop yard in Washington State. Eleven insect species or families showed significant attraction to 13 HIPVs. The ladybeetle, Stethorus punctum picipes, was attracted to sticky traps baited with methyl salicylate (MeSA), cis-3-hexen-1-ol (He), and benzaldehyde (Be). The minute pirate bug, Orius tristicolor, was attracted to traps baited with MeSA, He, Be, and octyl aldehyde (Oa), and the bigeyed bug, Geocoris pallens, responded to MeSA, indole, and trans-2-hexen-1-al. The mymarid wasp, Anagrus daanei, was attracted to He, Oa, and farnesene. The chloropid fly, Thaumatomyia glabra, was highly attracted to methyl anthranilate. Insect families responding to HIPVs included Syrphidae (MeSA, He), Braconidae ((Z)-3-hexenyl acetate, He, cis-jasmone (J), methyl jasmonate (MeJA), methyl anthranilate (MeA)), Empididae (MeSA), Sarcophagidae (MeSA, Be, J, nonanal and geraniol), Tachinidae (Be), and Agromyzidae (MeSA). Micro-Hymenoptera (primarily parasitic wasp families) were attracted to MeSA, He, and indole. These results are discussed with respect to known properties and bioactivity of the tested HIPVs and to their potential as tools for recruiting natural enemies into agroecosystems.
Sigma-Aldrich
cis-3-Hexen-1-ol, ≥98%, FCC, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
Geraniol, 98%