Merck
  • Pharmacokinetics of sublingual sufentanil tablets and efficacy and safety in the management of postoperative pain.
Pharmacokinetics of sublingual sufentanil tablets and efficacy and safety in the management of postoperative pain.
Regional anesthesia and pain medicine (2012-12-29)
Harold S Minkowitz, Neil K Singla, Mark A Evashenk, Stephen S Hwang, Yu-Kun Chiang, Lawrence G Hamel, Pamela P Palmer
ABSTRACT
A sublingual sufentanil tablet is being developed as a potential alternative to intravenous (IV) opioids for the management of postoperative pain. The objective of these studies was to evaluate the pharmacokinetics, efficacy, and safety of sublingual sufentanil tablets for postoperative pain management. The pharmacokinetics of sublingual sufentanil 10 and 80 µg were compared with IV sufentanil in 12 subjects in a phase 1 study. The safety and efficacy of sublingual sufentanil (5-15 µg) were evaluated in double-blind, randomized, placebo-controlled phase 2 studies in patients undergoing knee replacement surgery (n = 101) or open abdominal (ABD) surgery (n = 94). The primary efficacy measurement was the summed pain intensity difference compared with baseline over 12 hours (SPID-12). Sublingual sufentanil pharmacokinetics were dose proportional following single doses of 10 and 80 µg. Plasma half-time (time from peak plasma concentration to 50% of peak concentration) was 80 to 90 minutes for sublingual sufentanil compared with 15 minutes or less for IV sufentanil. In the phase 2 studies, greater SPID-12 scores (ie, lower pain intensity) compared with placebo were observed for sublingual sufentanil 15 µg in the knee replacement study (P < 0.05) and for 10 and 15 µg in the ABD study (P < 0.01). All doses of sublingual sufentanil were well tolerated, and the incidence of adverse events was similar between the sublingual sufentanil and placebo groups. Sufentanil formulated as a sublingual solid dosage form provides a duration of action that allows effective analgesia for postoperative patients in a medically supervised setting.
Product Description
Supelco
Sufentanil Citrate solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Sufentanil-D5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®