Merck
  • Automatic structure determination of regular polysaccharides based solely on NMR spectroscopy.
Automatic structure determination of regular polysaccharides based solely on NMR spectroscopy.
Biomacromolecules (2011-10-01)
Magnus Lundborg, Carolina Fontana, Göran Widmalm
ABSTRACT
The structural analysis of polysaccharides requires that the sugar components and their absolute configurations are determined. We here show that this can be performed based on NMR spectroscopy by utilizing butanolysis with (+)- and (-)-2-butanol that gives the corresponding 2-butyl glycosides with characteristic (1)H and (13)C NMR chemical shifts. The subsequent computer-assisted structural determination by CASPER can then be based solely on NMR data in a fully automatic way as shown and implemented herein. The method is additionally advantageous in that reference data only have to be prepared once and from a user's point of view only the unknown sample has to be derivatized for use in CASPER.
Sigma-Aldrich
Mixed Bed Resin, DNA grade, for molecular biology, BioReagent
Supelco
TMD-8 hydrogen and hydroxide form, hydrogen and hydroxide form