Merck
  • Determination of nitrofurantoin, furazolidone and furaltadone in milk by high-performance liquid chromatography with electrochemical detection.
Determination of nitrofurantoin, furazolidone and furaltadone in milk by high-performance liquid chromatography with electrochemical detection.
Journal of chromatography. A (1997-03-14)
T Galeano Díaz, A Guiberteau Cabanillas, M I Acedo Valenzuela, C A Correa, F Salinas
ABSTRACT
A HPLC method with coulometric detection has been established to carry out the separation of the three nitrofuran derivatives, nitrofurantoin, furazolidone and furaltadone. A Nova-Pak C18 column (150 x 3.9 mm) and a Coulochem II detector from ESA have been used. After obtaining the hydrodynamic curves of the three compounds in the porous graphite electrode a potential of -600 mV was selected as the working potential. The influence of other variables such as mobile phase composition and flow-rate were studied. The mobile phase considered as an optimum was acetonitrile-0.1 M aqueous solution of sodium perchlorate (28:72), with 0.5% glacial acetic acid. The oxygen of the mobile phase was removed with a vacuum system on-line and a nitrogen stream was used to remove the oxygen of the samples. The calibration graphs and the detection limits were established. The method proposed was used, with good results, for the determination of the three compounds in milk.
Sigma-Aldrich
Furaltadone
Pricing and availability is not currently available.
Supelco
Furaltadone, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Pricing and availability is not currently available.