Merck
  • Propionic acid-metabisulphite-Schiff reagent for quick Feulgen staining.
Propionic acid-metabisulphite-Schiff reagent for quick Feulgen staining.
Microscopica acta (1981-05-01)
M K Dutt
ABSTRACT
This communication presents a method for the preparation of Schiff reagent in which N hydrochloric acid has been replaced by a low concentration of propionic acid. The result of using such a Schiff reagent indicates that the dye-reagent thus prepared is extrafast in action on acid-hydrolysed mammalian tissue sections. The intensity of nuclear colouration is also increased considerably, since the pH of the dye-reagent is 6.0. It is, therefore advocated that this newly developed Schiff reagent be used at 5 degrees C.
Sigma-Aldrich
Potassium disulfite, ≥98%
Sigma-Aldrich
Potassium disulfite, puriss., meets analytical specification of Ph.Eur., BP, NF, FCC, E224, 95.0-101.0% (iodometric), powder, 51.8-57.6% sulfur dioxide
Sigma-Aldrich
Potassium disulfite, BioUltra, ≥97.0% (RT)