Merck
  • Mass preparation of oligosaccharides by the hydrolysis of chondroitin sulfate polysaccharides with a subcritical water microreaction system.
Mass preparation of oligosaccharides by the hydrolysis of chondroitin sulfate polysaccharides with a subcritical water microreaction system.
Carbohydrate research (2013-03-05)
Shuhei Yamada, Keiichiro Matsushima, Haruo Ura, Nobuyuki Miyamoto, Kazuyuki Sugahara
ABSTRACT
The biological functions of chondroitin sulfate (CS) are executed by the interaction of specific oligosaccharide sequences in the polysaccharide chain with effective proteins. Thus, CS oligosaccharides are expected to have pharmacological applications. Furthermore, the demand for CS in health food supplements and medication is growing. However, the absorbency of CS polysaccharides in the digestive system is very low. Since the activity of orally administered CS is expected to increase by depolymerization, industrial production of CS oligosaccharides is required. In this study, hydrolysis with subcritical and super-critical water was applied to the depolymerization of CS for the first time, and hydrolytic conditions for oligosaccharide production were examined. CS oligosaccharides principally containing an N-acetyl-D-galactosamine residue at their reducing ends were successfully obtained. No significant desulfation was found in CS oligosaccharides prepared under optimized conditions. The production of CS oligosaccharides by this method will have a strong influence on the CS-related materials market.
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chondroitin 4-sulfate sodium salt from bovine trachea, mixture of isomers, main component: chondroitin 4-sulfate
Chondroitin sulfate sodium (marine), European Pharmacopoeia (EP) Reference Standard
Chondroitin sulfate sodium, European Pharmacopoeia (EP) Reference Standard