Merck
  • PML mediates glioblastoma resistance to mammalian target of rapamycin (mTOR)-targeted therapies.
PML mediates glioblastoma resistance to mammalian target of rapamycin (mTOR)-targeted therapies.
Proceedings of the National Academy of Sciences of the United States of America (2013-02-27)
Akio Iwanami, Beatrice Gini, Ciro Zanca, Tomoo Matsutani, Alvaro Assuncao, Ali Nael, Julie Dang, Huijun Yang, Shaojun Zhu, Jun Kohyama, Issay Kitabayashi, Webster K Cavenee, Timothy F Cloughesy, Frank B Furnari, Masaya Nakamura, Yoshiaki Toyama, Hideyuki Okano, Paul S Mischel
ABSTRACT
Despite their nearly universal activation of mammalian target of rapamycin (mTOR) signaling, glioblastomas (GBMs) are strikingly resistant to mTOR-targeted therapy. We analyzed GBM cell lines, patient-derived tumor cell cultures, and clinical samples from patients in phase 1 clinical trials, and find that the promyelocytic leukemia (PML) gene mediates resistance to mTOR-targeted therapies. Direct mTOR inhibitors and EGF receptor (EGFR) inhibitors that block downstream mTOR signaling promote nuclear PML expression in GBMs, and genetic overexpression and knockdown approaches demonstrate that PML prevents mTOR and EGFR inhibitor-dependent cell death. Low doses of the PML inhibitor, arsenic trioxide, abrogate PML expression and reverse mTOR kinase inhibitor resistance in vivo, thus markedly inhibiting tumor growth and promoting tumor cell death in mice. These results identify a unique role for PML in mTOR and EGFR inhibitor resistance and provide a strong rationale for a combination therapeutic strategy to overcome it.
Sigma-Aldrich
Arsenic(III) oxide, SAJ first grade, ≥99.0%
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)