Merck
  • Synthesis of complex benzenoids via the intermediate generation of o-benzynes through the hexadehydro-Diels-Alder reaction.
Synthesis of complex benzenoids via the intermediate generation of o-benzynes through the hexadehydro-Diels-Alder reaction.
Nature protocols (2013-02-16)
Beeraiah Baire, Dawen Niu, Patrick H Willoughby, Brian P Woods, Thomas R Hoye
ABSTRACT
The hexadehydro-Diels-Alder (HDDA) cascade enables the synthesis of complex benzenoid products with various substitution patterns through aryne intermediates. The first stage of this cascade involves the generation of a highly reactive ortho-benzyne intermediate by a net [4+2] cycloisomerization of a triyne substrate. The benzyne can be rapidly 'trapped' either intramolecularly or intermolecularly with myriad nucleophilic or π-bond-donating reactants. As a representative example of a general procedure for synthesizing highly substituted benzenoids, this protocol describes the synthesis of a typical triyne substrate and its use as the reactant in an HDDA cascade to form a phthalide. The synthetic procedure detailed herein (four chemical reactions) takes 16-20 h of active effort over a period of several days for the preparation of the triyne precursor and ∼2 h of active effort over a 3-d period for the generation and trapping of the benzyne and isolation of the phthalide product.
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Copper(I) Chloride ChemBeads, 15 wt% loading
Sigma-Aldrich
Phthalide, 98%
Sigma-Aldrich
Propargyl alcohol, 99%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%