Merck
  • Nano-size layered manganese-calcium oxide as an efficient and biomimetic catalyst for water oxidation under acidic conditions: comparable to platinum.
Nano-size layered manganese-calcium oxide as an efficient and biomimetic catalyst for water oxidation under acidic conditions: comparable to platinum.
Dalton transactions (Cambridge, England : 2003) (2013-02-12)
Mohammad Mahdi Najafpour, Kevin C Leonard, Fu-Ren F Fan, Mahmoud Amouzadeh Tabrizi, Allen J Bard, Cecil K King'ondu, Steven L Suib, Behzad Haghighi, Suleyman I Allakhverdiev
ABSTRACT
Inspired by Nature's catalyst, a nano-size layered manganese-calcium oxide showed a low overvoltage for water oxidation in acidic solutions, which is comparable to platinum.
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%
Sigma-Aldrich
Calcium oxide, reagent grade