Merck
  • Energy relaxation in CdSe nanocrystals: the effects of morphology and film preparation.
Energy relaxation in CdSe nanocrystals: the effects of morphology and film preparation.
Optics express (2013-02-15)
Bryan T Spann, Liangliang Chen, Xiulin Ruan, Xianfan Xu
ABSTRACT
Ultrafast time-resolved absorption spectroscopy is used to investigate exciton dynamics in CdSe nanocrystal films. The effects of morphology, quantum-dot versus quantum-rod, and preparation of nanocrystals in a thin film form are investigated. The measurements revealed longer intraband exciton relaxation in quantum-rods than in quantum-dots. The slowed relaxation in quantum-rods is due to mitigation of the Auger-relaxation mechanism from elongating the nanocrystal. In addition, the nanocrystal thin film showed long-lived confined acoustic phonons corresponding to the ellipsoidal breathing mode, contrary to others work on colloidal systems of CdSe nanocrystals.
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Pricing and availability is not currently available.