Merck
Anaerobic digestion of sunflower oil cake: a current overview.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-11-22)
M A De la Rubia, V Fernández-Cegrí, F Raposo, R Borja
ABSTRACT
Due to the chemical and physical structure of a lignocellulosic biomass, its anaerobic digestion (AD) is a slow and difficult process. In this paper, the results obtained from a batch biochemical methane potential (BMP) test and fed-batch mesophilic AD assays of sunflower oil cake (SuOC) are presented. Taking into account the low digestibility shown during one-stage experiments the methane yield decreased considerably after increasing the organic loading rate (OLR) from 2 to 3 g VS L(-1) d(-1), SuOC was subjected to a two-stage AD process (hydrolytic-acidogenic and methanogenic stages), in two separate reactors operating in series where the methanogenic stage became acidified (with >1,600 mg acetic acid L(-1)) at an OLR as low as 2 g VS L(-1) d(-1). More recently, BMP assays were carried out after mechanical, thermal, and ultrasonic pre-treatments to determine the best option on the basis of the methane yield obtained.
Sigma-Aldrich
Sunflower oil, Certified organic (NOP)
Supelco
Sunflower seed oil, analytical standard
Sigma-Aldrich
Sunflower seed oil from Helianthus annuus