Merck
  • Telescoping phenomenon in pathological gambling: association with gender and comorbidities.
Telescoping phenomenon in pathological gambling: association with gender and comorbidities.
The Journal of nervous and mental disease (2012-11-06)
Jon E Grant, Brian L Odlaug, Marc E Mooney
ABSTRACT
The course of pathological gambling (PG) in women has been described as having a later age of initiation but a shorter time to problematic gambling ("telescoped"). This study examined evidence for telescoping and its relationship with comorbidities. Seventy-one treatment-seeking individuals with PG underwent a diagnostic interview to examine gambling behaviors, age at initiation of gambling, and time from initiation to meeting criteria for PG. The women had a higher mean age at gambling initiation compared with that of the men (mean [SD] age, 31.3 [13.0] years, compared with 22.4 [7.9] years; p = 0.0003) and a significantly shorter time from initiation of gambling to meeting the criteria for PG (8.33 [8.7] years compared with 11.97 [9.1] years; p = 0.0476) after controlling for demographic and clinical variables. This study presents evidence for a gender-specific course of PG unrelated to psychiatric comorbidities and suggests a need for greater clinical focus on the gender differences of gambling behavior.
Sigma-Aldrich
Tolcapone, ≥98% (HPLC)