Merck
  • Intermolecular chiral assemblies in R(-) and S(+) 2-butanol detected by microcalorimetry measurements.
Intermolecular chiral assemblies in R(-) and S(+) 2-butanol detected by microcalorimetry measurements.
Chirality (2012-05-10)
Uri Cogan, Avi Shpigelman, Irina Portnaya, Abraham Rutenberg, Yosef Scolnik, Meir Shinitzky
ABSTRACT
Supramolecular chiral assemblies of R(-) and S(+) 2-butanol, in their neat form or when dissolved in their nonchiral isomer isobutanol, were evaluated by isothermal titration calorimetry (ITC) ensuing mixing. Dilution of 0.5 M solution of R(-) 2-butanol in isobutanol into the latter liberated heat of several calories per mole, which was approximately double than that obtained in parallel dilutions of S(+) 2-butanol in isobutanol. The ITC dilution profiles indicated an estimate of about 100 isobutanol solvent molecules surrounding each of the 2-butanol enantiomers, presumably arranged in chiral configurations, with different adopted order between the isomers. Mixings of neat R and S 2-butanol were followed by endothermic ITC profiles, indicating that, in racemic 2-butanol, both the supramolecular order and the intermolecular binding energies are lower than in each of the neat chiral isomers. The diversion from symmetrical ITC patterns in these mixings indicated again a subtle difference in molecular organization between the neat enantiomers. It should be noted that the presence of impurities, α-pinene and teterhydrofuran, at a level totaling 0.5%, did not influence the ITC heat flow profiles. The findings of this study demonstrate for the first time that chiral solutes in organic solvents are expected to acquire asymmetric solvent envelopes that may be different between the enantiomers, thus broadening this phenomenon beyond the previously demonstrated cases in aqueous solutions.
Sigma-Aldrich
Aluminum-tri-sec-butoxide, 97%
Supelco
2-Butanol, analytical standard
Sigma-Aldrich
2-Butanol, anhydrous, 99.5%
Sigma-Aldrich
2-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%
Sigma-Aldrich
(S)-(+)-2-Butanol, 99%