Merck
  • Dithizone modified magnetic nanoparticles for fast and selective solid phase extraction of trace elements in environmental and biological samples prior to their determination by ICP-OES.
Dithizone modified magnetic nanoparticles for fast and selective solid phase extraction of trace elements in environmental and biological samples prior to their determination by ICP-OES.
Talanta (2012-01-24)
Guihong Cheng, Man He, Hanyong Peng, Bin Hu
ABSTRACT
A fast and simple method for analysis of trace amounts of Cr(III), Cu(II), Pb(II) and Zn(II) in environmental and biological samples was developed by combining magnetic solid phase extraction (MSPE) with inductively coupled plasma-optical emission spectrometry (ICP-OES) detection. Dithizone modified silica-coated magnetic Fe(3)O(4) nanoparticles (H(2)Dz-SCMNPs) were prepared and used for MSPE of trace amounts of Cr(III), Cu(II), Pb(II) and Zn(II). The prepared magnetic nanoparticles were characterized by scanning electron microscopy (SEM), transmission electron microscopy (TEM), X-ray powder diffraction (XRD), and Fourier transform infrared spectroscopy (FT-IR). The factors affecting the extraction of the target metal ions such as pH, sample volume, eluent, and interfering ions had been investigated and the adsorption mechanism of the target metals on the self-prepared H(2)Dz-SCMNPs was investigated by FT-IR and X-ray photo electron spectroscopy (XPS). Under the optimized conditions, the detection limits of the developed method for Cr(III), Cu(II), Pb(II) and Zn(II) were 35, 11, 62, and 8ngL(-1), respectively, with the enrichment factor of 100. The relative standard deviations (RSDs, c=10μgL(-1), n=7) were in the range of 1.7-3.1% and the linear range was 0.1-100μgL(-1). The proposed method had been validated by two certified reference materials (GSBZ50009-88 environmental water and GBW07601 human hair), and the determined values were in good agreement with the certified values. The method was also applied for the determination of trace metals in real water and human hair samples with recoveries in the range of 85-110% for the spiked samples. The developed MSPE-ICP-OES method has the advantages of simplicity, rapidity, selectivity, high extraction efficiency and is suitable for the analysis of samples with large volume and complex matrix.
Sigma-Aldrich
Dithizone, JIS special grade, ≥85.0%
Sigma-Aldrich
Dithizone, ACS reagent, ≥85.0%
Sigma-Aldrich
Dithizone, Practical Grade
Sigma-Aldrich
Dithizone, ACS reagent, for spectrophotometric det. of Cd, Cu, Hg, Pb, Zn, ≥98.0% (TLC)