Merck
  • Speciation and monitoring test for inorganic arsenic in white rice flour.
Speciation and monitoring test for inorganic arsenic in white rice flour.
Journal of agricultural and food chemistry (2012-01-10)
Tomohiro Narukawa, Akiharu Hioki, Koichi Chiba
ABSTRACT
A monitoring test for arsenic species in white rice flour was developed and applied to flours made from 20 samples of polished rice collected from locations all over Japan. The arsenic species in white rice flour made from five samples each of four types of rice were analyzed by HPLC-ICP-MS after a heat-assisted aqueous extraction. The total arsenic and major and minor element concentrations in the white rice flours were measured by ICP-MS and ICP-OES after microwave-assisted digestion. 91 ± 1% of the arsenic in the flours was extractable. Concentrations of arsenite [As(III)], arsenate [As(V)], and dimethylarsinic acid (DMAA) were closely positively correlated with the total arsenic concentrations. The total arsenic concentration in flours made from rice collected around Japan was 0.15 ± 0.07 mg kg(-1) (highest, 0.32 mg kg(-1)), which is very low. It was thus confirmed that the white rice flour samples collected in this experiment were not suffered from noticeable As contamination.
Sigma-Aldrich
Cacodylic acid, ≥99.0%
Sigma-Aldrich
Cacodylic acid, ≥98%
Cacodylic acid, analytical standard