Merck
  • Solution-processed MoO₃ thin films as a hole-injection layer for organic solar cells.
Solution-processed MoO₃ thin films as a hole-injection layer for organic solar cells.
ACS applied materials & interfaces (2011-08-13)
Claudio Girotto, Eszter Voroshazi, David Cheyns, Paul Heremans, Barry P Rand
ABSTRACT
We report on a sol-gel-based technique to fabricate MoO(3) thin films as a hole-injection layer for solution-processed or thermally evaporated organic solar cells. The solution-processed MoO(3) (sMoO(3)) films are demonstrated to have equal performance to hole-injection layers composed of either PEDOT:PSS or thermally evaporated MoO(3) (eMoO(3)), and the annealing temperature at which the sol-gel layer begins to work is consistent with the thermodynamic analysis of the process. Finally, the shelf lifetime of devices made with the sMoO(3) is similar to equivalent devices prepared with a eMoO(3) hole-injection layer.
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%