Merck
  • Conversion of airborne nerolidol to DMNT emission requires additional signals in Achyranthes bidentata.
Conversion of airborne nerolidol to DMNT emission requires additional signals in Achyranthes bidentata.
FEBS letters (2011-04-23)
Shigeru Tamogami, Yukiko Takahashi, Makoto Abe, Koji Noge, Randeep Rakwal, Ganesh Kumar Agrawal
ABSTRACT
DMNT biosynthesis was proposed to proceed via (E)-nerolidol in plants a decade ago. However, (E)-nerolidol function as airborne signal/substrate for in-vivo biosynthesis of DMNT remains to be investigated and the regulation of DMNT production and emission is largely unknown. We address both of these aspects using Achyranthes bidentata model plant in conjunction with deuterium-labeled d(5)-(E)-nerolidol, headspace, GC-FID, and GC/MS-based absolute quantification approaches. We demonstrate that airborne (E)-nerolidol is specifically metabolized in-vivo into DMNT emission, but requires airborne VOC MeJA or predator herbivore as additional environmental signal. In addition, we provide new insight into the complex regulation underlying DMNT emission, and highlight the importance of studying multiple environmental factors on emission patterns of plant VOCs and their mechanistic regulation.
Product Description
Supelco
trans-Nerolidol, analytical standard
Sigma-Aldrich
cis-Nerolidol, ≥96.0% (GC)
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans