Merck
  • Tolerance to 1-pentene-3-ol and to 1-pentene-3-one in relation to alcohol dehydrogenase (ADH) and aldo keto reductase (AKR) activities in Drosophila melanogaster.
Tolerance to 1-pentene-3-ol and to 1-pentene-3-one in relation to alcohol dehydrogenase (ADH) and aldo keto reductase (AKR) activities in Drosophila melanogaster.
Biochemical genetics (1990-10-01)
J J Garrido, M Barbancho
ABSTRACT
The detoxification of 1-pentene-3-ol (pentenol) and 1-pentene-3-one (pentenone) by Drosophila melanogaster adult flies has been studied in two homozygous lines for the AdhF and AdhS alleles (LRC lines), in their respective lines selected for tolerance to ethanol (LRSe lines) and in a homozygous strain for the Adhn4 null allele. For each line, the genotype and sex LDs50 of both compounds were estimated. Then, in order to explain the differences in LD50, both alcohol dehydrogenase (ADH) and aldo keto reductase (AKR) activities were assayed. In addition, the effects of pentenone on AKR activity were also studied. Our results show that ADH-positive flies exhibit a much higher sensitivity to pentenol than ADH-null flies. However, both ADH-positive and ADH-null flies show a similar tolerance to pentenone. Our results show that flies selected for improving tolerance to ethanol also have increased tolerance to pentenol (FF and SS flies) and pentenone (SS flies). However, this improved ability to tolerate pentenol and/or pentenone cannot be explained by changes in ADH or AKR activities. On the other hand, we have observed a beneficial effect of pentenol, but not of pentenone, in n4 flies. We also show that AKR activity is not modified by the administration of pentenone. These results suggest that, in the absence of ADH activity, pentenol may be transformed into a compound that is less toxic than pentenone and that pentenone itself might also be transformed into a less toxic compound.
Sigma-Aldrich
Ethyl vinyl ketone, ≥97%, stabilized with BHT, FG
Sigma-Aldrich
1-Penten-3-one, contains 0.1% BHT as stabilizer, 97%
Sigma-Aldrich
1-Penten-3-ol, 99%
Sigma-Aldrich
1-Penten-3-ol, ≥98%, FG